Search
Search
| Gene Symbol: Gorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100563.1 | NP_001094033.1 | 2 - 3 | 490 | 65 | |
| XM_008769650.1 | XP_008767872.1 | 1 - 2 | 1066 | 65 | ||
| GenBank mRNA | BC160825.1 | - | 2 - 3 | 490 | 65 | |
Set Membership: |
|