Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01484594_m1
          See other FSHB GE Assays ›
          Gene Symbol:
          Fshb
          Gene Name:
          follicle stimulating hormone, beta polypeptide
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 98088321 - 98093144 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01484594_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fshb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007597.1 NP_001007598.1 1 - 2 155 73
          XM_006234672.2 XP_006234734.1 2 - 3 1230 73
          GenBank mRNA BC168724.1 - 2 - 3 217 6889701 73
          M36804.1 - 1 - 2 168 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003821.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fshb

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          transforming growth factor beta receptor signaling pathway
          follicle-stimulating hormone signaling pathway
          regulation of osteoclast differentiation
          positive regulation of bone resorption
          Sertoli cell proliferation
          follicle-stimulating hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560111 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079887 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079887 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079887 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15830 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848012 - -
          Affymetrix Rat Gene 1.0 ST Array 10848009 - -
          Affymetrix Rat Gene 1.0 ST Array 10848011 - -
          Affymetrix Rat Gene 1.0 ST Array 10848010 - -
          Affymetrix Rat Gene 1.1 ST Array 10848008 - -
          Rat Genome 230 2.0 Array 1388289_PM_s_at - -
          Rat Genome 230 2.0 Array 1388094_PM_at - -
          Rat Genome 230 2.0 Array 1388289_s_at - -
          Rat Genome 230 2.0 Array 1388094_at - -
          Affymetrix HT Rat Focus Array 1388094_at - -
          Affymetrix HT Rat Focus Array 1388289_PM_s_at - -
          Affymetrix HT Rat Focus Array 1388094_PM_at - -
          Affymetrix HT Rat Focus Array 1388289_s_at - -
          Affymetrix Rat Clariom S Assay PSR0300029152.rn.1 TC0300003821.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300029153.rn.1 TC0300003821.rn.1 RTA TC
          Rat Expression Set 230 1388094_PM_at - -
          Rat Expression Set 230 1388094_at - -
          Rat Expression Set 230 1388289_s_at - -
          Rat Expression Set 230 1388289_PM_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848008 - -
          Rat Genome U34 Set M36804_s_at - -
          Rat Genome U34 Set M27048_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22393203 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline