Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01484657_m1
          See other CYBRD1 GE Assays ›
          Gene Symbol:
          Cybrd1
          Gene Name:
          cytochrome b reductase 1
          Gene Aliases:
          Dcytb
          Chromosome Location:
          Chr.3: 57718139 - 57745749 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01484657_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cybrd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011954.1 NP_001011954.1 3 - 4 557 65
          XM_008761891.1 XP_008760113.1 2 - 3 705 65
          GenBank mRNA BC085768.1 - 3 - 4 557 7108043 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000794.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cybrd1

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to iron ion
          oxidation-reduction process
          ferric-chelate reductase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P084088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 197645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166817 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101747 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109995 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084088 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P084088 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P084088 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46295 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102810 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43454 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836791 - -
          Affymetrix Rat Gene 1.0 ST Array 10846127 - -
          Affymetrix Rat Gene 1.0 ST Array 10836792 - -
          Affymetrix Rat Gene 1.0 ST Array 10836793 - -
          Affymetrix Rat Gene 1.0 ST Array 10836790 - -
          Affymetrix Rat Gene 1.1 ST Array 10846126 - -
          Affymetrix Rat Gene 1.1 ST Array 10836788 - -
          Rat Genome 230 2.0 Array 1377369_at - -
          Rat Genome 230 2.0 Array 1377369_PM_at - -
          Affymetrix HT Rat Focus Array 1377369_at - -
          Affymetrix HT Rat Focus Array 1377369_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300005988.rn.1 TC0300000794.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005989.rn.1 TC0300000794.rn.1 RTA TC
          Rat Expression Set 230 1377369_at - -
          Rat Expression Set 230 1377369_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10836788 - -
          Rat Genome U34 Set AB007601mRNA_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21154635 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375782 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0