Search
Search

Zfyve19| Gene Symbol: Zfyve19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034948.1 | NP_001030120.1 | 1 - 2 | 143 | 58 | |
| XM_006234786.2 | XP_006234848.1 | 1 - 2 | 79 | 58 | ||
| XM_006234787.2 | XP_006234849.1 | 1 - 2 | 79 | 58 | ||
| XM_008762106.1 | XP_008760328.1 | 1 - 2 | 143 | 58 | ||
| GenBank mRNA | BC099795.1 | - | 1 - 2 | 143 | 7936116 | 58 |
Set Membership: |
|