Search
Search
| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013119.1 | NP_037251.1 | 13 - 14 | 2413 | 69 | |
| XM_008761914.1 | XP_008760136.1 | 12 - 13 | 2312 | 69 | ||
| XM_008761915.1 | XP_008760137.1 | 13 - 14 | 2315 | 69 | ||
| XM_008761916.1 | XP_008760138.1 | 12 - 13 | 2313 | 69 | ||
| XM_008761917.1 | XP_008760139.1 | 12 - 13 | 2312 | 69 | ||
| XM_008761918.1 | XP_008760140.1 | 12 - 13 | 2225 | 69 | ||
| XM_008761919.1 | XP_008760141.1 | 12 - 13 | 2216 | 69 | ||
| XM_008761920.1 | XP_008760142.1 | 12 - 13 | 2216 | 69 | ||
| GenBank mRNA | Y00766.1 | - | 13 - 14 | 2413 | 69 | |
Set Membership: |
|