Search
Search

Dync1i2| Gene Symbol: Dync1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270624.1 | NP_001257553.1 | 6 - 7 | 852 | 63 | |
| NM_001270625.1 | NP_001257554.1 | 5 - 6 | 792 | 63 | ||
| NM_053880.2 | NP_446332.2 | 7 - 8 | 870 | 63 | ||
| XM_006234344.2 | XP_006234406.1 | 6 - 7 | 629 | 63 | ||
| XM_006234346.2 | XP_006234408.1 | 5 - 6 | 568 | 63 | ||
| GenBank mRNA | BC078764.1 | - | 6 - 7 | 598 | 7113840 | 63 |
| U39044.1 | - | 7 - 8 | 586 | 63 | ||
| U39045.1 | - | 6 - 7 | 568 | 63 | ||
| U39046.1 | - | 5 - 6 | 508 | 63 | ||
Set Membership: |
|