Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01485600_m1
          See other SERPING1 GE Assays ›
          Gene Symbol:
          Serping1
          Gene Name:
          serpin peptidase inhibitor, clade G (C1 inhibitor), member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 72161230 - 72171109 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01485600_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serping1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199093.1 NP_954524.1 6 - 7 1313 83
          XM_006234447.2 XP_006234509.1 7 - 8 1388 83
          GenBank mRNA BC061860.1 - 6 - 7 1313 5598525 83
          FQ219227.1 - 5 - 6 1018 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003442.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serping1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of complement activation, lectin pathway
          complement activation, classical pathway
          aging
          blood coagulation
          biological_process
          negative regulation of endopeptidase activity
          fibrinolysis
          innate immune response
          complement binding
          molecular_function
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P080025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90623 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51913 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080025 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P080025 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P080025 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P510386 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P422630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43477 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846860 - -
          Affymetrix Rat Gene 1.0 ST Array 10846858 - -
          Affymetrix Rat Gene 1.0 ST Array 10846856 - -
          Affymetrix Rat Gene 1.0 ST Array 10846857 - -
          Affymetrix Rat Gene 1.0 ST Array 10846861 - -
          Affymetrix Rat Gene 1.0 ST Array 10846859 - -
          Affymetrix Rat Gene 1.0 ST Array 10846855 - -
          Affymetrix Rat Gene 1.1 ST Array 10846854 - -
          Rat Genome 230 2.0 Array 1372254_PM_at - -
          Rat Genome 230 2.0 Array 1372254_at - -
          Affymetrix HT Rat Focus Array 1372254_PM_at - -
          Affymetrix HT Rat Focus Array 1372254_at - -
          Affymetrix Rat Clariom S Assay PSR0300027134.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027141.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027142.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027143.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027140.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027136.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027138.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027139.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027137.rn.1 TC0300003442.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027135.rn.1 TC0300003442.rn.1 RTA TC
          Rat Expression Set 230 1372254_PM_at - -
          Rat Expression Set 230 1372254_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846854 - -
          Rat Genome U34 Set rc_AA800318_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21255586 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374314 - -
          RatRef-12 Expression BeadChip ILMN_60046 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline