Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01485715_g1
          See other NOP56 GE Assays ›
          Gene Symbol:
          Nop56
          Gene Name:
          NOP56 ribonucleoprotein
          Gene Aliases:
          Nol5a
          Chromosome Location:
          Chr.3: 122803728 - 122808564 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01485715_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nop56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025732.1 NP_001020903.1 8 - 9 1065 71
          GenBank mRNA BC086568.1 - 5 - 6 743 71
          BC099149.1 - 8 - 9 1065 7366316 71
          FQ232640.1 - 3 - 4 311 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001711.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nop56

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA modification
          snoRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000596 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P160571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363394 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160571 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P160571 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P160571 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000596 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P842946 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1029770 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19558 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839858 - -
          Affymetrix Rat Gene 1.0 ST Array 10839865 - -
          Affymetrix Rat Gene 1.0 ST Array 10839864 - -
          Affymetrix Rat Gene 1.0 ST Array 10839862 - -
          Affymetrix Rat Gene 1.0 ST Array 10839859 - -
          Affymetrix Rat Gene 1.0 ST Array 10839861 - -
          Affymetrix Rat Gene 1.0 ST Array 10839860 - -
          Affymetrix Rat Gene 1.0 ST Array 10839866 - -
          Affymetrix Rat Gene 1.0 ST Array 10839863 - -
          Affymetrix Rat Gene 1.0 ST Array 10839867 - -
          Affymetrix Rat Gene 1.1 ST Array 10839857 - -
          Rat Genome 230 2.0 Array 1395998_at - -
          Rat Genome 230 2.0 Array 1388622_at - -
          Rat Genome 230 2.0 Array 1388622_PM_at - -
          Rat Genome 230 2.0 Array 1395998_PM_at - -
          Affymetrix HT Rat Focus Array 1388622_PM_at - -
          Affymetrix HT Rat Focus Array 1395998_at - -
          Affymetrix HT Rat Focus Array 1388622_at - -
          Affymetrix HT Rat Focus Array 1395998_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300012280.rn.1 TC0300001711.rn.1 RTA TC
          Rat Expression Set 230 1388622_at - -
          Rat Expression Set 230 1388622_PM_at - -
          Rat Expression Set 230 1395998_at - -
          Rat Expression Set 230 1395998_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839857 - -
          Rat Genome U34 Set rc_AA924405_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22148391 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355261 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline