Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01485897_m1
          See other MPPED2 GE Assays ›
          Gene Symbol:
          Mpped2
          Gene Name:
          metallophosphoesterase domain containing 2
          Gene Aliases:
          C11orf8, C11orf8h
          Chromosome Location:
          Chr.3: 97720308 - 97899735 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01485897_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mpped2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198778.2 NP_942073.2 5 - 6 802 63
          XM_006234680.2 XP_006234742.1 5 - 6 2177 63
          XM_006234681.2 XP_006234743.1 5 - 6 879 63
          XM_006234682.2 XP_006234744.1 5 - 6 966 63
          XM_006234683.2 XP_006234745.1 5 - 6 881 63
          XM_008762104.1 XP_008760326.1 5 - 6 1601 63
          XM_008762105.1 XP_008760327.1 4 - 5 1861 63
          GenBank mRNA AY093422.1 - 5 - 6 852 63
          BC161828.1 - 5 - 6 884 9033590 63
          EV768250.1 - 3 - 4 437 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001347.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mpped2

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P219695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125739 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549064 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119882 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100211 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95090 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130745 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59802 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41064 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54475 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48011 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56171 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219695 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P219695 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P219695 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P219695 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48954 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8909 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96462 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78535 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9351 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79700 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104512 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29959 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68366 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1142 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76248 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42667 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10838313 - -
          Affymetrix Rat Gene 1.0 ST Array 10838315 - -
          Affymetrix Rat Gene 1.0 ST Array 10838317 - -
          Affymetrix Rat Gene 1.0 ST Array 10838316 - -
          Affymetrix Rat Gene 1.0 ST Array 10838314 - -
          Affymetrix Rat Gene 1.0 ST Array 10838319 - -
          Affymetrix Rat Gene 1.0 ST Array 10838318 - -
          Affymetrix Rat Gene 1.1 ST Array 10838312 - -
          Rat Genome 230 2.0 Array 1392077_PM_at - -
          Rat Genome 230 2.0 Array 1392077_at - -
          Affymetrix HT Rat Focus Array 1392077_at - -
          Affymetrix HT Rat Focus Array 1392077_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300009138.rn.1 TC0300001347.rn.1 RTA TC
          Rat Expression Set 230 1392077_at - -
          Rat Expression Set 230 1392077_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10838312 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22030622 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373221 - -
          RatRef-12 Expression BeadChip ILMN_56703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline