Search
Search
| Gene Symbol: Ddrgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106512.1 | NP_001099982.1 | 2 - 3 | 726 | 60 | |
| GenBank mRNA | FQ214225.1 | - | 2 - 3 | 311 | 60 | |
| FQ231424.1 | - | 2 - 3 | 350 | 60 | ||
Set Membership: |
|