Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01486277_m1
          See other DPH6 GE Assays ›
          Gene Symbol:
          Dph6
          Gene Name:
          diphthamine biosynthesis 6
          Gene Aliases:
          Atpbd4, RGD1310006
          Chromosome Location:
          Chr.3: 106007721 - 106136662 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01486277_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dph6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014181.1 NP_001014203.1 7 - 8 789 70
          GenBank mRNA BC087148.1 - 7 - 8 789 7131040 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003922.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dph6

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-diphthamide biosynthetic process from peptidyl-histidine
          ATP binding
          diphthine-ammonia ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81179 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268266 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53396 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111753 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149739 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60116 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56718 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84646 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63913 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26620 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59609 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119062 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119062 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119062 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28236 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61273 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92642 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93174 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81487 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49821 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35303 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848367 - -
          Affymetrix Rat Gene 1.0 ST Array 10848365 - -
          Affymetrix Rat Gene 1.0 ST Array 10848366 - -
          Affymetrix Rat Gene 1.0 ST Array 10848363 - -
          Affymetrix Rat Gene 1.0 ST Array 10848369 - -
          Affymetrix Rat Gene 1.0 ST Array 10848364 - -
          Affymetrix Rat Gene 1.0 ST Array 10848368 - -
          Affymetrix Rat Gene 1.1 ST Array 10848362 - -
          Rat Genome 230 2.0 Array 1389708_PM_at - -
          Rat Genome 230 2.0 Array 1389708_at - -
          Affymetrix HT Rat Focus Array 1389708_PM_at - -
          Affymetrix HT Rat Focus Array 1389708_at - -
          Affymetrix Rat Clariom S Assay PSR0300029723.rn.1 TC0300003922.rn.1 RTA TC
          Rat Expression Set 230 1389708_PM_at - -
          Rat Expression Set 230 1389708_at - -
          Affymetrix Rat Gene 1.0 ST Array 10848362 - -
          Rat Genome U34 Set rc_AA900481_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21993634 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649794 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline