Search
Search
| Gene Symbol: Cep295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242549.2 | XP_006242611.2 | 3 - 4 | 547 | 61 | |
| XM_008765949.1 | XP_008764171.1 | 3 - 4 | 547 | 61 | ||
| XM_008765950.1 | XP_008764172.1 | 3 - 4 | 547 | 61 | ||
| XM_008765951.1 | XP_008764173.1 | 3 - 4 | 547 | 61 | ||
| XM_008765952.1 | XP_008764174.1 | 3 - 4 | 547 | 61 | ||
| XM_008765953.1 | XP_008764175.1 | 3 - 4 | 547 | 61 | ||
| XM_008765954.1 | XP_008764176.1 | 3 - 4 | 548 | 61 | ||
| XM_008765955.1 | XP_008764177.1 | 3 - 4 | 547 | 61 | ||
| XM_008765956.1 | XP_008764178.1 | 3 - 4 | 547 | 61 | ||
| XM_008765957.1 | XP_008764179.1 | 3 - 4 | 547 | 61 | ||
Set Membership: |
|