Search
Search

Slc44a2| Gene Symbol: Slc44a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134715.1 | NP_001128187.1 | 14 - 15 | 1306 | 65 | |
| XM_006242649.2 | XP_006242711.1 | 14 - 15 | 2056 | 65 | ||
| XM_006242650.2 | XP_006242712.1 | 14 - 15 | 1326 | 65 | ||
| XM_006242651.2 | XP_006242713.1 | 14 - 15 | 1326 | 65 | ||
| GenBank mRNA | BC168183.1 | - | 14 - 15 | 1306 | 7098013 | 65 |
| FQ225791.1 | - | 14 - 15 | 1327 | 65 | ||
| FQ231292.1 | - | 14 - 15 | 1327 | 65 | ||
Set Membership: |
|