Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01487138_m1
          See other ENKUR GE Assays ›
          Gene Symbol:
          Enkur
          Gene Name:
          enkurin, TRPC channel interacting protein
          Gene Aliases:
          RGD1309839
          Chromosome Location:
          Chr.17: 88071794 - 88095444 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01487138_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Enkur
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106126.1 NP_001099596.1 4 - 5 807 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700002347.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P426717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123237 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P426717 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P426717 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P426717 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P426717 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66996 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10799978 - -
          Affymetrix Rat Gene 1.0 ST Array 10799982 - -
          Affymetrix Rat Gene 1.0 ST Array 10799979 - -
          Affymetrix Rat Gene 1.0 ST Array 10799980 - -
          Affymetrix Rat Gene 1.0 ST Array 10799981 - -
          Affymetrix Rat Gene 1.1 ST Array 10799977 - -
          Affymetrix Rat Clariom S Assay PSR1700015692.rn.1 TC1700002347.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700015693.rn.1 TC1700002347.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10799977 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22168752 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350223 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline