Search
Search
| Gene Symbol: Stox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134863.1 | NP_001128335.1 | 2 - 3 | 1675 | 86 | |
| XM_006253151.2 | XP_006253213.2 | 2 - 3 | 958 | 86 | ||
| XM_006253152.2 | XP_006253214.1 | 2 - 3 | 386 | 86 | ||
| XM_006253153.2 | XP_006253215.1 | 2 - 3 | 766 | 86 | ||
| XM_008771258.1 | XP_008769480.1 | 2 - 3 | 526 | 86 | ||
Set Membership: |
|