Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01488126_m1
          See other UFSP2 GE Assays ›
          Gene Symbol:
          Ufsp2
          Gene Name:
          UFM1-specific peptidase 2
          Gene Aliases:
          RGD1311161
          Chromosome Location:
          Chr.16: 49465958 - 49484975 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01488126_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ufsp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014142.1 NP_001014164.1 11 - 12 1363 104
          XM_006253159.2 XP_006253221.1 11 - 12 1353 104
          XM_008771265.1 XP_008769487.1 11 - 12 1514 104
          GenBank mRNA BC083771.1 - 11 - 12 1363 7193395 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001657.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ufsp2

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          regulation of intracellular estrogen receptor signaling pathway
          thiolester hydrolase activity
          UFM1 hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P286087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 493582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142804 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105478 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P286087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P286087 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P286087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P286087 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75714 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10791731 - -
          Affymetrix Rat Gene 1.0 ST Array 10791721 - -
          Affymetrix Rat Gene 1.0 ST Array 10791726 - -
          Affymetrix Rat Gene 1.0 ST Array 10791729 - -
          Affymetrix Rat Gene 1.0 ST Array 10791730 - -
          Affymetrix Rat Gene 1.0 ST Array 10791725 - -
          Affymetrix Rat Gene 1.0 ST Array 10791727 - -
          Affymetrix Rat Gene 1.0 ST Array 10791724 - -
          Affymetrix Rat Gene 1.0 ST Array 10791723 - -
          Affymetrix Rat Gene 1.0 ST Array 10791728 - -
          Affymetrix Rat Gene 1.0 ST Array 10791722 - -
          Affymetrix Rat Gene 1.0 ST Array 10791720 - -
          Affymetrix Rat Gene 1.1 ST Array 10791719 - -
          Rat Genome 230 2.0 Array 1376086_at - -
          Rat Genome 230 2.0 Array 1376086_PM_at - -
          Affymetrix HT Rat Focus Array 1376086_at - -
          Affymetrix HT Rat Focus Array 1376086_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600012385.rn.1 TC1600001657.rn.1 RTA TC
          Rat Expression Set 230 1376086_PM_at - -
          Rat Expression Set 230 1376086_at - -
          Affymetrix Rat Gene 1.0 ST Array 10791719 - -
          Rat Genome U34 Set rc_H33149_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21657808 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370603 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline