Search
Search

RGD1564308| Gene Symbol: RGD1564308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253169.2 | XP_006253231.1 | 5 - 6 | 896 | 69 | |
| XM_006253170.2 | XP_006253232.1 | 5 - 6 | 681 | 69 | ||
| XM_577523.6 | XP_577523.4 | 5 - 6 | 912 | 69 | ||
Set Membership: |
|