Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01488161_m1
          See other KLKB1 GE Assays ›
          Gene Symbol:
          Klkb1
          Gene Name:
          kallikrein B, plasma 1
          Gene Aliases:
          KALP15, Kalp1, Klk3
          Chromosome Location:
          Chr.16: 50151128 - 50175407 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01488161_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klkb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012725.2 NP_036857.2 14 - 15 1787 79
          XM_006253121.2 XP_006253183.1 14 - 15 2682 79
          XM_008771252.1 XP_008769474.1 12 - 13 1542 79
          XM_008771253.1 XP_008769475.1 12 - 13 1636 79
          GenBank mRNA BC089815.1 - 14 - 15 1787 7369524 79
          M30282.1 - 14 - 15 1750 79
          M58590.1 - 14 - 15 1750 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000595.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klkb1

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          inflammatory response
          blood coagulation
          biological_process
          plasminogen activation
          fibrinolysis
          positive regulation of fibrinolysis
          liver regeneration
          molecular_function
          serine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801760 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77419 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11534 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11534 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11534 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40525 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84413 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10788284 - -
          Affymetrix Rat Gene 1.0 ST Array 10788283 - -
          Affymetrix Rat Gene 1.0 ST Array 10788280 - -
          Affymetrix Rat Gene 1.0 ST Array 10788288 - -
          Affymetrix Rat Gene 1.0 ST Array 10788290 - -
          Affymetrix Rat Gene 1.0 ST Array 10788282 - -
          Affymetrix Rat Gene 1.0 ST Array 10788289 - -
          Affymetrix Rat Gene 1.0 ST Array 10788279 - -
          Affymetrix Rat Gene 1.0 ST Array 10788286 - -
          Affymetrix Rat Gene 1.0 ST Array 10788291 - -
          Affymetrix Rat Gene 1.0 ST Array 10788287 - -
          Affymetrix Rat Gene 1.0 ST Array 10788281 - -
          Affymetrix Rat Gene 1.0 ST Array 10788292 - -
          Affymetrix Rat Gene 1.0 ST Array 10788285 - -
          Affymetrix Rat Gene 1.1 ST Array 10788278 - -
          Rat Genome 230 2.0 Array 1387323_at - -
          Rat Genome 230 2.0 Array 1387323_PM_at - -
          Affymetrix HT Rat Focus Array 1387323_at - -
          Affymetrix HT Rat Focus Array 1387323_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600004794.rn.1 TC1600000595.rn.1 RTA TC
          Rat Expression Set 230 1387323_at - -
          Rat Expression Set 230 1387323_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10788278 - -
          Rat Genome U34 Set M30282_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20998578 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361123 - -
          RatRef-12 Expression BeadChip ILMN_58443 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline