Search
Search

Trappc11| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169115.1 | NP_001162586.1 | 19 - 20 | 2324 | 64 | |
| XM_006253134.2 | XP_006253196.1 | 19 - 20 | 2245 | 64 | ||
| XM_006253135.2 | XP_006253197.1 | 19 - 20 | 2180 | 64 | ||
Set Membership: |
|