Search
Search
| Gene Symbol: Slc7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134686.2 | NP_001128158.1 | 3 - 4 | 714 | 64 | |
| NM_022619.3 | NP_072141.2 | 3 - 4 | 714 | 64 | ||
| XM_008771281.1 | XP_008769503.1 | 3 - 4 | 5793 | 64 | ||
| XM_008771282.1 | XP_008769504.1 | 4 - 5 | 673 | 64 | ||
| XM_008771283.1 | XP_008769505.1 | 3 - 4 | 5793 | 64 | ||
| GenBank mRNA | AF245001.1 | - | 3 - 4 | 715 | 64 | |
| AF245002.1 | - | 3 - 4 | 715 | 64 | ||
Set Membership: |
|