Search
Search
| Gene Symbol: N6amt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134990.1 | NP_001128462.1 | 2 - 3 | 330 | 103 | |
| XM_006252059.2 | XP_006252121.1 | 3 - 4 | 1183 | 103 | ||
| XM_006252061.2 | XP_006252123.1 | 2 - 3 | 1011 | 103 | ||
| GenBank mRNA | BC168660.1 | - | 2 - 3 | 330 | 5621479 | 103 |
| FQ230419.1 | - | 3 - 4 | 380 | 103 | ||
Set Membership: |
|