Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01489190_m1
          See other TM9SF1 GE Assays ›
          Gene Symbol:
          Tm9sf1
          Gene Name:
          transmembrane 9 superfamily member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 34314645 - 34322390 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01489190_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tm9sf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012155.1 NP_001012155.1 3 - 4 1529 69
          XM_006251990.2 XP_006252052.1 4 - 5 1369 69
          XM_006251991.2 XP_006252053.1 4 - 5 1385 69
          XM_008770688.1 XP_008768910.1 3 - 4 1586 69
          GenBank mRNA BC081891.1 - 3 - 4 1529 7109579 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001714.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tm9sf1

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s)

          autophagy

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P618274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P618274 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P618274 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P618274 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P618274 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18031 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10783853 - -
          Affymetrix Rat Gene 1.0 ST Array 10783854 - -
          Affymetrix Rat Gene 1.0 ST Array 10783852 - -
          Affymetrix Rat Gene 1.0 ST Array 10783850 - -
          Affymetrix Rat Gene 1.0 ST Array 10783855 - -
          Affymetrix Rat Gene 1.1 ST Array 10783849 - -
          Rat Genome 230 2.0 Array 1390125_at - -
          Rat Genome 230 2.0 Array 1390125_PM_at - -
          Affymetrix HT Rat Focus Array 1390125_PM_at - -
          Affymetrix HT Rat Focus Array 1390125_at - -
          Affymetrix Rat Clariom S Assay PSR1500011112.rn.1 TC1500001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011105.rn.1 TC1500001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011110.rn.1 TC1500001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011107.rn.1 TC1500001714.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500011108.rn.1 TC1500001714.rn.1 RTA TC
          Rat Expression Set 230 1390125_at - -
          Rat Expression Set 230 1390125_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10783849 - -
          Rat Genome U34 Set rc_AA964064_at - -
          Rat Genome U34 Set rc_AI176626_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21526600 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376407 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline