Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01489315_m1
See other ITGA2 GE Assays ›
Gene Symbol:
Itga2
Gene Name:
integrin, alpha 2
Gene Aliases:
CD49B
Chromosome Location:
Chr.2: 46996904 - 47097011 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01489315_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Itga2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_345156.7
XP_345157.5
14 - 15
1973
52
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Rattus norvegicus
Inventoried
Probe spans exons
Panther Classification:
Molecular Function -
integrin
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of leukocyte migration
movement of cell or subcellular component
substrate-dependent cell migration
hypotonic response
cell adhesion
cell-matrix adhesion
integrin-mediated signaling pathway
female pregnancy
cell proliferation
positive regulation of epithelial cell migration
positive regulation of alkaline phosphatase activity
response to organic cyclic compound
response to amine
response to muscle activity
positive regulation of smooth muscle cell migration
mammary gland development
positive regulation of cell projection organization
cell-substrate adhesion
positive regulation of collagen biosynthetic process
positive regulation of collagen binding
response to L-ascorbic acid
cell adhesion mediated by integrin
collagen-activated signaling pathway
wound healing
response to drug
positive regulation of DNA binding
skin morphogenesis
establishment of protein localization
positive regulation of translation
positive regulation of cell adhesion
positive regulation of smooth muscle contraction
focal adhesion assembly
mesodermal cell differentiation
positive regulation of smooth muscle cell proliferation
positive regulation of inflammatory response
positive regulation of positive chemotaxis
detection of mechanical stimulus involved in sensory perception of pain
positive regulation of transmission of nerve impulse
positive regulation of phagocytosis, engulfment
hepatocyte differentiation
response to parathyroid hormone
cellular response to mechanical stimulus
cellular response to estradiol stimulus
cellular response to organic cyclic compound
integrin binding
collagen binding
protein complex binding
collagen receptor activity
laminin binding
protein heterodimerization activity
cell adhesion molecule binding
collagen binding involved in cell-matrix adhesion
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline