Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01489493_m1
          See other RGD1306227 GE Assays ›
          Gene Symbol:
          RGD1306227
          Gene Name:
          similar to 4833420G17Rik protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 52333396 - 52355044 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01489493_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1306227
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014037.1 NP_001014059.1 12 - 13 1908 72
          GenBank mRNA BC078991.1 - 12 - 13 1908 7112582 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000465.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1306227

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124192 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21709 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138071 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019798 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019798 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019798 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P470980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85231 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47293 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10813196 - -
          Affymetrix Rat Gene 1.0 ST Array 10813197 - -
          Affymetrix Rat Gene 1.0 ST Array 10813203 - -
          Affymetrix Rat Gene 1.0 ST Array 10813195 - -
          Affymetrix Rat Gene 1.0 ST Array 10813200 - -
          Affymetrix Rat Gene 1.0 ST Array 10813199 - -
          Affymetrix Rat Gene 1.0 ST Array 10813198 - -
          Affymetrix Rat Gene 1.0 ST Array 10813201 - -
          Affymetrix Rat Gene 1.0 ST Array 10813205 - -
          Affymetrix Rat Gene 1.0 ST Array 10813202 - -
          Affymetrix Rat Gene 1.0 ST Array 10813193 - -
          Affymetrix Rat Gene 1.0 ST Array 10813192 - -
          Affymetrix Rat Gene 1.0 ST Array 10813204 - -
          Affymetrix Rat Gene 1.0 ST Array 10813194 - -
          Affymetrix Rat Gene 1.1 ST Array 10813191 - -
          Affymetrix Rat Clariom S Assay PSR0200002671.rn.1 TC0200000465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002672.rn.1 TC0200000465.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200002673.rn.1 TC0200000465.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10813191 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21967983 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369676 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline