Search
Search

Ankrd32| Gene Symbol: Ankrd32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106400.1 | NP_001099870.1 | 15 - 16 | 2091 | 106 | |
| XM_008760599.1 | XP_008758821.1 | 13 - 14 | 1733 | 106 | ||
Set Membership: |
|