Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01489894_m1
          See other RAD17 GE Assays ›
          Gene Symbol:
          Rad17
          Gene Name:
          RAD17 homolog (S. pombe)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 30634353 - 30664628 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01489894_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rad17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024778.1 NP_001019949.1 14 - 15 1819 86
          XM_006231848.2 XP_006231910.1 14 - 15 1815 86
          GenBank mRNA BC098057.1 - 14 - 15 1819 7457767 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002739.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rad17

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          DNA repair
          cellular response to DNA damage stimulus
          mitotic cell cycle checkpoint
          negative regulation of DNA replication
          regulation of phosphorylation
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P208464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460552 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73616 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174403 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114546 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P208464 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P208464 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P208464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P208464 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26335 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62513 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10820990 - -
          Affymetrix Rat Gene 1.0 ST Array 10820981 - -
          Affymetrix Rat Gene 1.0 ST Array 10820982 - -
          Affymetrix Rat Gene 1.0 ST Array 10820976 - -
          Affymetrix Rat Gene 1.0 ST Array 10820977 - -
          Affymetrix Rat Gene 1.0 ST Array 10820989 - -
          Affymetrix Rat Gene 1.0 ST Array 10820978 - -
          Affymetrix Rat Gene 1.0 ST Array 10820974 - -
          Affymetrix Rat Gene 1.0 ST Array 10820988 - -
          Affymetrix Rat Gene 1.0 ST Array 10820987 - -
          Affymetrix Rat Gene 1.0 ST Array 10820983 - -
          Affymetrix Rat Gene 1.0 ST Array 10820985 - -
          Affymetrix Rat Gene 1.0 ST Array 10820984 - -
          Affymetrix Rat Gene 1.0 ST Array 10820986 - -
          Affymetrix Rat Gene 1.0 ST Array 10820980 - -
          Affymetrix Rat Gene 1.0 ST Array 10820975 - -
          Affymetrix Rat Gene 1.0 ST Array 10820979 - -
          Affymetrix Rat Gene 1.1 ST Array 10820973 - -
          Rat Genome 230 2.0 Array 1385090_at - -
          Rat Genome 230 2.0 Array 1385090_PM_at - -
          Affymetrix HT Rat Focus Array 1385090_at - -
          Affymetrix HT Rat Focus Array 1385090_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200019321.rn.1 TC0200002739.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200019318.rn.1 TC0200002739.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200019319.rn.1 TC0200002739.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200019320.rn.1 TC0200002739.rn.1 RTA TC
          Rat Expression Set 230 1385090_at - -
          Rat Expression Set 230 1385090_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10820973 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22403336 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350980 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline