Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01489978_m1
          See other NR2F1 GE Assays ›
          Gene Symbol:
          Nr2f1
          Gene Name:
          nuclear receptor subfamily 2, group F, member 1
          Gene Aliases:
          COUP-TFI
          Chromosome Location:
          Chr.2: 5569954 - 5579894 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01489978_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr2f1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031130.1 NP_112392.1 1 - 2 930 78
          GenBank mRNA U10995.1 - 1 - 2 930 78
          U15002.1 - 1 - 2 116 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002509.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr2f1

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neuron migration
          transcription, DNA-templated
          nervous system development
          positive regulation of receptor biosynthetic process
          negative regulation of neuron projection development
          cerebral cortex regionalization
          intracellular receptor signaling pathway
          forebrain development
          steroid hormone mediated signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          zinc ion binding
          sequence-specific DNA binding
          retinoic acid-responsive element binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P138636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73518 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112783 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88333 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138636 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P138636 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P138636 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14310 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P541011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10820129 - -
          Affymetrix Rat Gene 1.0 ST Array 10820130 - -
          Affymetrix Rat Gene 1.0 ST Array 10820128 - -
          Affymetrix Rat Gene 1.0 ST Array 10820127 - -
          Affymetrix Rat Gene 1.0 ST Array 10820131 - -
          Affymetrix Rat Gene 1.1 ST Array 10820126 - -
          Rat Genome 230 2.0 Array 1387571_PM_at - -
          Rat Genome 230 2.0 Array 1387571_at - -
          Affymetrix HT Rat Focus Array 1387571_at - -
          Affymetrix HT Rat Focus Array 1387571_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200017600.rn.1 TC0200002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017593.rn.1 TC0200002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017598.rn.1 TC0200002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017597.rn.1 TC0200002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017599.rn.1 TC0200002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017592.rn.1 TC0200002509.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200017594.rn.1 TC0200002509.rn.1 RTA TC
          Rat Expression Set 230 1387571_PM_at - -
          Rat Expression Set 230 1387571_at - -
          Affymetrix Rat Gene 1.0 ST Array 10820126 - -
          Rat Genome U34 Set U10995_at - -
          Rat Genome U34 Set U10995_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21336932 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1649840 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline