Search
Search
| Gene Symbol: Lax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017491.1 | NP_001017491.1 | 3 - 4 | 538 | 111 | |
| XM_006249793.2 | XP_006249855.1 | 3 - 4 | 673 | 111 | ||
| GenBank mRNA | BC089837.1 | - | 3 - 4 | 538 | 7365271 | 111 |
Set Membership: |
|