Search
Search
| Gene Symbol: Pou2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100639.1 | NP_001094109.1 | 14 - 15 | 1908 | 58 | |
| XM_008769591.1 | XP_008767813.1 | 14 - 15 | 4034 | 58 | ||
| XM_008769592.1 | XP_008767814.1 | 14 - 15 | 2169 | 58 | ||
| XM_008769593.1 | XP_008767815.1 | 16 - 17 | 2368 | 58 | ||
| XM_008769594.1 | XP_008767816.1 | 15 - 16 | 2270 | 58 | ||
| XM_008769595.1 | XP_008767817.1 | 16 - 17 | 2303 | 58 | ||
| XM_008769596.1 | XP_008767818.1 | 16 - 17 | 4478 | 58 | ||
| XM_008769597.1 | XP_008767819.1 | 15 - 16 | 4380 | 58 | ||
| XM_008769598.1 | XP_008767820.1 | 14 - 15 | 4279 | 58 | ||
| XM_008769599.1 | XP_008767821.1 | 14 - 15 | 1932 | 58 | ||
| GenBank mRNA | U17013.1 | - | 9 - 10 | 1498 | 58 | |
Set Membership: |
|