Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490556_m1
          See other TSEN15 GE Assays ›
          Gene Symbol:
          Tsen15
          Gene Name:
          TSEN15 tRNA splicing endonuclease subunit
          Gene Aliases:
          RGD1308584
          Chromosome Location:
          Chr.13: 69765093 - 69781019 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490556_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsen15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105958.1 NP_001099428.1 1 - 2 228 93
          XM_006249984.2 XP_006250046.1 1 - 2 223 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001629.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA splicing, via endonucleolytic cleavage and ligation
          RNA phosphodiester bond hydrolysis, endonucleolytic
          tRNA-intron endonuclease activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1045724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86855 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53716 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1045724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52052 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768585 - -
          Affymetrix Rat Gene 1.0 ST Array 10768587 - -
          Affymetrix Rat Gene 1.0 ST Array 10768586 - -
          Affymetrix Rat Gene 1.1 ST Array 10768583 - -
          Rat Genome 230 2.0 Array 1373848_at - -
          Rat Genome 230 2.0 Array 1373848_PM_at - -
          Affymetrix HT Rat Focus Array 1373848_at - -
          Affymetrix HT Rat Focus Array 1373848_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300011127.rn.1 TC1300001629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011129.rn.1 TC1300001629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011131.rn.1 TC1300001629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011126.rn.1 TC1300001629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011132.rn.1 TC1300001629.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300011130.rn.1 TC1300001629.rn.1 RTA TC
          Rat Expression Set 230 1373848_PM_at - -
          Rat Expression Set 230 1373848_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768583 - -
          Rat Genome U34 Set rc_AI045154_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352015 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline