Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490562_m1
          See other XPR1 GE Assays ›
          Gene Symbol:
          Xpr1
          Gene Name:
          xenotropic and polytropic retrovirus receptor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 72913377 - 73056785 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490562_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xpr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105992.1 NP_001099462.1 14 - 15 2122 105
          XM_008769626.1 XP_008767848.1 14 - 15 3011 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001680.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          response to virus
          receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P029847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122307 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138902 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17959 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39906 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121820 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116572 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3962 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63539 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128170 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029847 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P029847 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P029847 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P339936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60469 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76476 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74739 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768835 - -
          Affymetrix Rat Gene 1.0 ST Array 10768849 - -
          Affymetrix Rat Gene 1.0 ST Array 10768836 - -
          Affymetrix Rat Gene 1.0 ST Array 10768839 - -
          Affymetrix Rat Gene 1.0 ST Array 10768843 - -
          Affymetrix Rat Gene 1.0 ST Array 10768841 - -
          Affymetrix Rat Gene 1.0 ST Array 10768846 - -
          Affymetrix Rat Gene 1.0 ST Array 10768840 - -
          Affymetrix Rat Gene 1.0 ST Array 10768838 - -
          Affymetrix Rat Gene 1.0 ST Array 10768848 - -
          Affymetrix Rat Gene 1.0 ST Array 10768845 - -
          Affymetrix Rat Gene 1.0 ST Array 10768844 - -
          Affymetrix Rat Gene 1.0 ST Array 10768850 - -
          Affymetrix Rat Gene 1.0 ST Array 10768842 - -
          Affymetrix Rat Gene 1.0 ST Array 10768837 - -
          Affymetrix Rat Gene 1.1 ST Array 10768834 - -
          Rat Genome 230 2.0 Array 1375846_PM_at - -
          Rat Genome 230 2.0 Array 1375846_at - -
          Affymetrix HT Rat Focus Array 1375846_PM_at - -
          Affymetrix HT Rat Focus Array 1375846_at - -
          Affymetrix Rat Clariom S Assay PSR1300011589.rn.1 TC1300001680.rn.1 RTA TC
          Rat Expression Set 230 1375846_PM_at - -
          Rat Expression Set 230 1375846_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768834 - -
          Rat Genome U34 Set rc_AI058281_at - -
          Rat Genome U34 Set rc_AI113099_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22086605 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357833 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline