Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490608_m1
          See other CHI3L1 GE Assays ›
          Gene Symbol:
          Chi3l1
          Gene Name:
          chitinase 3-like 1 (cartilage glycoprotein-39)
          Gene Aliases:
          Chil1
          Chromosome Location:
          Chr.13: 51022826 - 51030797 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490608_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chi3l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053560.1 NP_446012.1 6 - 7 637 107
          XM_006249892.2 XP_006249954.2 6 - 7 691 107
          XM_008769574.1 XP_008767796.1 6 - 7 658 107
          GenBank mRNA AF062038.1 - 4 - 5 518 107
          BC091365.1 - 6 - 7 637 7319171 107
          EF467168.1 - 4 - 5 518 107
          FQ232458.1 - 3 - 4 303 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000303.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chi3l1

          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          chitin catabolic process
          apoptotic process
          inflammatory response
          immune response
          activation of NF-kappaB-inducing kinase activity
          response to mechanical stimulus
          positive regulation of peptidyl-threonine phosphorylation
          lung development
          response to tumor necrosis factor
          positive regulation of angiogenesis
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          response to interleukin-6
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          interleukin-8 secretion
          molecular_function
          chitinase activity
          chitin binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P452176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921217 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36376 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452176 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452176 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P452176 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452176 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701440290 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56348 - -
          Agilent Rat cDNA Microarray (G4105A) 701440290 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10764071 - -
          Affymetrix Rat Gene 1.0 ST Array 10764077 - -
          Affymetrix Rat Gene 1.0 ST Array 10764070 - -
          Affymetrix Rat Gene 1.0 ST Array 10764078 - -
          Affymetrix Rat Gene 1.0 ST Array 10764073 - -
          Affymetrix Rat Gene 1.0 ST Array 10764075 - -
          Affymetrix Rat Gene 1.0 ST Array 10764076 - -
          Affymetrix Rat Gene 1.0 ST Array 10764072 - -
          Affymetrix Rat Gene 1.0 ST Array 10764074 - -
          Affymetrix Rat Gene 1.1 ST Array 10764069 - -
          Rat Genome 230 2.0 Array 1392171_PM_at - -
          Rat Genome 230 2.0 Array 1392171_at - -
          Affymetrix HT Rat Focus Array 1392171_at - -
          Affymetrix HT Rat Focus Array 1392171_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1300001922.rn.1 TC1300000303.rn.1 RTA TC
          Rat Expression Set 230 1392171_PM_at - -
          Rat Expression Set 230 1392171_at - -
          Affymetrix Rat Gene 1.0 ST Array 10764069 - -
          Rat Genome U34 Set rc_AA945643_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22221549 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359657 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline