Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01490784_m1
          See other MDM4 GE Assays ›
          Gene Symbol:
          Mdm4
          Gene Name:
          MDM4, p53 regulator
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 49786776 - 49828786 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01490784_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mdm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012026.1 NP_001012026.1 7 - 8 629 122
          XM_006249777.2 XP_006249839.1 7 - 8 690 122
          XM_006249778.2 XP_006249840.1 7 - 8 694 122
          XM_006249779.2 XP_006249841.1 7 - 8 691 122
          GenBank mRNA BC083647.1 - 7 - 8 629 7135053 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001411.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mdm4

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein complex assembly
          biological_process
          cell proliferation
          positive regulation of cell proliferation
          DNA damage response, signal transduction by p53 class mediator
          negative regulation of protein catabolic process
          negative regulation of apoptotic process
          G0 to G1 transition
          protein stabilization
          negative regulation of cell cycle arrest
          cellular response to hypoxia
          molecular_function
          zinc ion binding
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P17202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 723840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17202 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P17202 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P17202 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P17202 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20580 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28231 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45515 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87487 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95123 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767669 - -
          Affymetrix Rat Gene 1.0 ST Array 10767667 - -
          Affymetrix Rat Gene 1.0 ST Array 10767672 - -
          Affymetrix Rat Gene 1.0 ST Array 10767673 - -
          Affymetrix Rat Gene 1.0 ST Array 10767665 - -
          Affymetrix Rat Gene 1.0 ST Array 10767664 - -
          Affymetrix Rat Gene 1.0 ST Array 10767674 - -
          Affymetrix Rat Gene 1.0 ST Array 10767671 - -
          Affymetrix Rat Gene 1.0 ST Array 10767668 - -
          Affymetrix Rat Gene 1.0 ST Array 10767670 - -
          Affymetrix Rat Gene 1.0 ST Array 10767666 - -
          Affymetrix Rat Gene 1.1 ST Array 10767663 - -
          Rat Genome 230 2.0 Array 1382417_PM_at - -
          Rat Genome 230 2.0 Array 1382417_at - -
          Affymetrix HT Rat Focus Array 1382417_PM_at - -
          Affymetrix HT Rat Focus Array 1382417_at - -
          Affymetrix Rat Clariom S Assay PSR1300009433.rn.1 TC1300001411.rn.1 RTA TC
          Rat Expression Set 230 1382417_PM_at - -
          Rat Expression Set 230 1382417_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767663 - -
          Rat Genome U34 Set rc_AI179906_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20723139 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376455 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0