Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01491003_m1
          See other NFASC GE Assays ›
          Gene Symbol:
          Nfasc
          Gene Name:
          neurofascin
          Gene Aliases:
          NF
          Chromosome Location:
          Chr.13: 49335407 - 49522470 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01491003_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nfasc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001160313.1 NP_001153785.1 2 - 3 289 92
          NM_001160314.1 NP_001153786.1 5 - 6 525 92
          NM_001160315.1 NP_001153787.1 4 - 5 716 92
          NM_053909.2 NP_446361.1 4 - 5 507 92
          XM_006249727.2 XP_006249789.1 5 - 6 580 92
          XM_006249734.2 XP_006249796.1 5 - 6 579 92
          XM_006249738.2 XP_006249800.1 4 - 5 560 92
          XM_006249741.2 XP_006249803.1 5 - 6 501 92
          XM_006249742.2 XP_006249804.1 5 - 6 450 92
          XM_008769406.1 XP_008767628.1 5 - 6 705 92
          XM_008769407.1 XP_008767629.1 4 - 5 687 92
          XM_008769408.1 XP_008767630.1 5 - 6 705 92
          XM_008769409.1 XP_008767631.1 5 - 6 705 92
          XM_008769410.1 XP_008767632.1 4 - 5 687 92
          XM_008769411.1 XP_008767633.1 5 - 6 705 92
          XM_008769412.1 XP_008767634.1 5 - 6 705 92
          XM_008769413.1 XP_008767635.1 5 - 6 705 92
          XM_008769414.1 XP_008767636.1 5 - 6 704 92
          XM_008769415.1 XP_008767637.1 5 - 6 704 92
          XM_008769416.1 XP_008767638.1 4 - 5 686 92
          XM_008769417.1 XP_008767639.1 4 - 5 686 92
          XM_008769418.1 XP_008767640.1 4 - 5 686 92
          XM_008769419.1 XP_008767641.1 4 - 5 686 92
          GenBank mRNA AY061639.1 - 2 - 3 289 92
          L11002.1 - 1256 92
          U81035.1 - 2 - 3 127 92
          U81036.1 - 3 - 4 145 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001405.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nfasc

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization to paranode region of axon
          cell adhesion
          axon guidance
          peripheral nervous system development
          transmission of nerve impulse
          paranodal junction assembly
          heterotypic cell-cell adhesion
          myelination
          clustering of voltage-gated sodium channels
          synapse organization
          protein localization to juxtaparanode region of axon
          protein targeting to plasma membrane
          protein domain specific binding
          protein binding involved in heterotypic cell-cell adhesion

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143629 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P108896 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P727294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546120 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339279 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54994 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136551 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97708 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10379 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161888 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115650 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42429 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P727294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143629 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108896 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P727294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108896 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143629 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P108896 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143629 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P727294 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P196467 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700518412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49457 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29216 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98090 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32226 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19143 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35866 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71728 - -
          Agilent Rat cDNA Microarray (G4105A) 700518412 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10767661 - -
          Affymetrix Rat Gene 1.0 ST Array 10767642 - -
          Affymetrix Rat Gene 1.0 ST Array 10767657 - -
          Affymetrix Rat Gene 1.0 ST Array 10767653 - -
          Affymetrix Rat Gene 1.0 ST Array 10767662 - -
          Affymetrix Rat Gene 1.0 ST Array 10767649 - -
          Affymetrix Rat Gene 1.0 ST Array 10767658 - -
          Affymetrix Rat Gene 1.0 ST Array 10767644 - -
          Affymetrix Rat Gene 1.0 ST Array 10767645 - -
          Affymetrix Rat Gene 1.0 ST Array 10767647 - -
          Affymetrix Rat Gene 1.0 ST Array 10767646 - -
          Affymetrix Rat Gene 1.0 ST Array 10767637 - -
          Affymetrix Rat Gene 1.0 ST Array 10767639 - -
          Affymetrix Rat Gene 1.0 ST Array 10767654 - -
          Affymetrix Rat Gene 1.0 ST Array 10767638 - -
          Affymetrix Rat Gene 1.0 ST Array 10767640 - -
          Affymetrix Rat Gene 1.0 ST Array 10767634 - -
          Affymetrix Rat Gene 1.0 ST Array 10767632 - -
          Affymetrix Rat Gene 1.0 ST Array 10767633 - -
          Affymetrix Rat Gene 1.0 ST Array 10767635 - -
          Affymetrix Rat Gene 1.0 ST Array 10767650 - -
          Affymetrix Rat Gene 1.0 ST Array 10767651 - -
          Affymetrix Rat Gene 1.0 ST Array 10767648 - -
          Affymetrix Rat Gene 1.0 ST Array 10767641 - -
          Affymetrix Rat Gene 1.0 ST Array 10767652 - -
          Affymetrix Rat Gene 1.0 ST Array 10767660 - -
          Affymetrix Rat Gene 1.0 ST Array 10767656 - -
          Affymetrix Rat Gene 1.0 ST Array 10767659 - -
          Affymetrix Rat Gene 1.0 ST Array 10767643 - -
          Affymetrix Rat Gene 1.0 ST Array 10767636 - -
          Affymetrix Rat Gene 1.1 ST Array 10767631 - -
          Rat Genome 230 2.0 Array 1388041_s_at - -
          Rat Genome 230 2.0 Array 1388041_PM_s_at - -
          Rat Genome 230 2.0 Array 1370877_PM_at - -
          Rat Genome 230 2.0 Array 1370876_at - -
          Rat Genome 230 2.0 Array 1374164_at - -
          Rat Genome 230 2.0 Array 1374164_PM_at - -
          Rat Genome 230 2.0 Array 1380917_PM_at - -
          Rat Genome 230 2.0 Array 1393181_at - -
          Rat Genome 230 2.0 Array 1370876_PM_at - -
          Rat Genome 230 2.0 Array 1393181_PM_at - -
          Rat Genome 230 2.0 Array 1370877_at - -
          Rat Genome 230 2.0 Array 1380917_at - -
          Affymetrix HT Rat Focus Array 1370877_at - -
          Affymetrix HT Rat Focus Array 1370877_PM_at - -
          Affymetrix HT Rat Focus Array 1388041_PM_s_at - -
          Affymetrix HT Rat Focus Array 1388041_s_at - -
          Affymetrix Rat Clariom S Assay PSR1300009384.rn.1 TC1300001405.rn.1 RTA TC
          Rat Expression Set 230 1374164_at - -
          Rat Expression Set 230 1370877_at - -
          Rat Expression Set 230 1374164_PM_at - -
          Rat Expression Set 230 1388041_s_at - -
          Rat Expression Set 230 1388041_PM_s_at - -
          Rat Expression Set 230 1370877_PM_at - -
          Rat Expression Set 230 1393181_at - -
          Rat Expression Set 230 1380917_at - -
          Rat Expression Set 230 1393181_PM_at - -
          Rat Expression Set 230 1380917_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10767631 - -
          Rat Genome U34 Set U81035_s_at - -
          Rat Genome U34 Set U81036_s_at - -
          Rat Genome U34 Set L11002_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368040 - -
          RatRef-12 Expression BeadChip ILMN_67353 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0