Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01491285_g1
          See other SRFBP1 GE Assays ›
          Gene Symbol:
          Srfbp1
          Gene Name:
          serum response factor binding protein 1
          Gene Aliases:
          p49/STRAP
          Chromosome Location:
          Chr.18: 47456406 - 47478532 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01491285_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srfbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001005536.1 NP_001005536.1 7 - 8 1201 152
          XM_008772109.1 XP_008770331.1 5 - 6 1082 152
          GenBank mRNA BC082077.1 - 7 - 8 1201 7133229 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000507.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srfbp1

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          maturation of SSU-rRNA
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P496934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102077 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90594 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P496934 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P496934 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P496934 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P496934 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80044 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14129 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68847 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801703 - -
          Affymetrix Rat Gene 1.0 ST Array 10801705 - -
          Affymetrix Rat Gene 1.0 ST Array 10801706 - -
          Affymetrix Rat Gene 1.0 ST Array 10801700 - -
          Affymetrix Rat Gene 1.0 ST Array 10801702 - -
          Affymetrix Rat Gene 1.0 ST Array 10801704 - -
          Affymetrix Rat Gene 1.0 ST Array 10801699 - -
          Affymetrix Rat Gene 1.0 ST Array 10801701 - -
          Affymetrix Rat Gene 1.1 ST Array 10801698 - -
          Rat Genome 230 2.0 Array 1378221_at - -
          Rat Genome 230 2.0 Array 1377827_at - -
          Rat Genome 230 2.0 Array 1378221_PM_at - -
          Rat Genome 230 2.0 Array 1377827_PM_at - -
          Affymetrix HT Rat Focus Array 1378221_at - -
          Affymetrix HT Rat Focus Array 1378221_PM_at - -
          Affymetrix HT Rat Focus Array 1377827_PM_at - -
          Affymetrix HT Rat Focus Array 1377827_at - -
          Affymetrix Rat Clariom S Assay PSR1800003700.rn.1 TC1800000507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800003701.rn.1 TC1800000507.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800003699.rn.1 TC1800000507.rn.1 RTA TC
          Rat Expression Set 230 1377827_at - -
          Rat Expression Set 230 1377827_PM_at - -
          Rat Expression Set 230 1378221_PM_at - -
          Rat Expression Set 230 1378221_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801698 - -
          Rat Genome U34 Set rc_AI171338_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22233885 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351300 - -
          RatRef-12 Expression BeadChip ILMN_64983 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline