Search
Search
| Gene Symbol: Spire1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107381.1 | NP_001100851.1 | 13 - 14 | 1915 | 71 | |
| XM_006254860.2 | XP_006254922.1 | 15 - 16 | 2187 | 71 | ||
| XM_006254862.2 | XP_006254924.1 | 14 - 15 | 2145 | 71 | ||
| XM_006254863.2 | XP_006254925.1 | 14 - 15 | 2012 | 71 | ||
| XM_006254864.2 | XP_006254926.1 | 13 - 14 | 1970 | 71 | ||
Set Membership: |
|