Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc6a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053996.2 | NP_446448.2 | 1048 | 124 | ||
GenBank mRNA | M88111.1 | - | 1048 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |