Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01491716_m1
          See other ZFP532 GE Assays ›
          Gene Symbol:
          Zfp532
          Gene Name:
          zinc finger protein 532
          Gene Aliases:
          Znf532
          Chromosome Location:
          Chr.18: 61257522 - 61368371 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01491716_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp532
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107382.1 NP_001100852.1 8 - 9 3416 88
          XM_006254865.2 XP_006254927.1 8 - 9 5706 88
          XM_006254866.2 XP_006254928.1 8 - 9 5145 88
          XM_006254868.2 XP_006254930.1 8 - 9 3733 88
          XM_006254869.2 XP_006254931.1 8 - 9 3578 88
          XM_006254870.2 XP_006254932.1 8 - 9 5110 88
          XM_006254871.2 XP_006254933.1 7 - 8 3291 88
          XM_006254872.2 XP_006254934.1 8 - 9 4490 88
          XM_006254873.2 XP_006254935.1 7 - 8 5596 88
          XM_006254875.2 XP_006254937.1 4 - 5 3810 88
          XM_006254876.2 XP_006254938.1 3 - 4 3528 88
          XM_008772142.1 XP_008770364.1 7 - 8 4332 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000699.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P545059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607336 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395916 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619468 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88070 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165667 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163157 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60337 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74332 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P545059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P545059 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P545059 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P297609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85890 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76627 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802302 - -
          Affymetrix Rat Gene 1.0 ST Array 10802301 - -
          Affymetrix Rat Gene 1.0 ST Array 10802300 - -
          Affymetrix Rat Gene 1.0 ST Array 10802305 - -
          Affymetrix Rat Gene 1.0 ST Array 10802307 - -
          Affymetrix Rat Gene 1.0 ST Array 10802306 - -
          Affymetrix Rat Gene 1.0 ST Array 10802296 - -
          Affymetrix Rat Gene 1.0 ST Array 10802304 - -
          Affymetrix Rat Gene 1.0 ST Array 10802299 - -
          Affymetrix Rat Gene 1.0 ST Array 10802298 - -
          Affymetrix Rat Gene 1.0 ST Array 10802297 - -
          Affymetrix Rat Gene 1.0 ST Array 10802303 - -
          Affymetrix Rat Gene 1.1 ST Array 10802294 - -
          Rat Genome 230 2.0 Array 1379800_PM_at - -
          Rat Genome 230 2.0 Array 1375792_at - -
          Rat Genome 230 2.0 Array 1375792_PM_at - -
          Rat Genome 230 2.0 Array 1379800_at - -
          Rat Genome 230 2.0 Array 1391478_PM_at - -
          Rat Genome 230 2.0 Array 1391478_at - -
          Affymetrix HT Rat Focus Array 1391478_at - -
          Affymetrix HT Rat Focus Array 1391478_PM_at - -
          Affymetrix HT Rat Focus Array 1379800_PM_at - -
          Affymetrix HT Rat Focus Array 1375792_at - -
          Affymetrix HT Rat Focus Array 1379800_at - -
          Affymetrix HT Rat Focus Array 1375792_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800005113.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005093.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005100.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005108.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005111.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005101.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005103.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005109.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005099.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005106.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005107.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005112.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005096.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005104.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005102.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005105.rn.1 TC1800000699.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800005110.rn.1 TC1800000699.rn.1 RTA TC
          Rat Expression Set 230 1375792_at - -
          Rat Expression Set 230 1375792_PM_at - -
          Rat Expression Set 230 1391478_PM_at - -
          Rat Expression Set 230 1379800_at - -
          Rat Expression Set 230 1379800_PM_at - -
          Rat Expression Set 230 1391478_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802294 - -
          Rat Genome U34 Set rc_AA819654_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22348251 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360861 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline