Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01491742_m1
          See other LOC361346 GE Assays ›
          Gene Symbol:
          LOC361346
          Gene Name:
          similar to chromosome 18 open reading frame 54
          Gene Aliases:
          Las2
          Chromosome Location:
          Chr.18: 68957469 - 68983253 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01491742_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC361346
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017462.1 NP_001017462.1 8 - 9 1455 127
          XM_006254929.2 XP_006254991.1 6 - 7 1285 127
          XM_008772154.1 XP_008770376.1 6 - 7 1528 127
          GenBank mRNA BC082051.1 - 8 - 9 1455 7132090 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800001675.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC361346

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P546117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19433 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171648 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P546117 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P546117 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P546117 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P546117 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65497 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805102 - -
          Affymetrix Rat Gene 1.0 ST Array 10805110 - -
          Affymetrix Rat Gene 1.0 ST Array 10805103 - -
          Affymetrix Rat Gene 1.0 ST Array 10805109 - -
          Affymetrix Rat Gene 1.0 ST Array 10805107 - -
          Affymetrix Rat Gene 1.0 ST Array 10805101 - -
          Affymetrix Rat Gene 1.0 ST Array 10805104 - -
          Affymetrix Rat Gene 1.0 ST Array 10805105 - -
          Affymetrix Rat Gene 1.0 ST Array 10805108 - -
          Affymetrix Rat Gene 1.1 ST Array 10805100 - -
          Rat Genome 230 2.0 Array 1379740_at - -
          Rat Genome 230 2.0 Array 1380293_PM_at - -
          Rat Genome 230 2.0 Array 1380293_at - -
          Rat Genome 230 2.0 Array 1379740_PM_at - -
          Affymetrix HT Rat Focus Array 1380293_PM_at - -
          Affymetrix HT Rat Focus Array 1380293_at - -
          Affymetrix HT Rat Focus Array 1379740_PM_at - -
          Affymetrix HT Rat Focus Array 1379740_at - -
          Affymetrix Rat Clariom S Assay PSR1800011742.rn.1 TC1800001675.rn.1 RTA TC
          Rat Expression Set 230 1379740_PM_at - -
          Rat Expression Set 230 1379740_at - -
          Rat Expression Set 230 1380293_at - -
          Rat Expression Set 230 1380293_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10805100 - -
          Rat Genome U34 Set rc_AI639413_at - -
          Rat Genome U34 Set rc_AI175283_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22323381 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352522 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline