Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01491927_s1
          See other CHMP1B GE Assays ›
          Gene Symbol:
          Chmp1b
          Gene Name:
          charged multivesicular body protein 1B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 62923620 - 62926150 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01491927_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chmp1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109533.1 NP_001103003.1 1 2150 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000725.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell separation after cytokinesis
          nucleus organization
          mitotic metaphase plate congression
          regulation of centrosome duplication
          viral budding via host ESCRT complex
          establishment of protein localization
          regulation of mitotic spindle assembly
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167737 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011719 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011719 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011719 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802406 - -
          Affymetrix Rat Gene 1.1 ST Array 10802405 - -
          Rat Genome 230 2.0 Array 1385158_at - -
          Rat Genome 230 2.0 Array 1390128_PM_at - -
          Rat Genome 230 2.0 Array 1390128_at - -
          Rat Genome 230 2.0 Array 1382129_PM_at - -
          Rat Genome 230 2.0 Array 1382112_PM_at - -
          Rat Genome 230 2.0 Array 1385158_PM_at - -
          Rat Genome 230 2.0 Array 1382112_at - -
          Rat Genome 230 2.0 Array 1382129_at - -
          Affymetrix HT Rat Focus Array 1382112_PM_at - -
          Affymetrix HT Rat Focus Array 1382112_at - -
          Affymetrix HT Rat Focus Array 1385158_PM_at - -
          Affymetrix HT Rat Focus Array 1385158_at - -
          Affymetrix HT Rat Focus Array 1382129_at - -
          Affymetrix HT Rat Focus Array 1382129_PM_at - -
          Affymetrix HT Rat Focus Array 1390128_at - -
          Affymetrix HT Rat Focus Array 1390128_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800005222.rn.1 TC1800000725.rn.1 RTA TC
          Rat Expression Set 230 1390128_PM_at - -
          Rat Expression Set 230 1390128_at - -
          Rat Expression Set 230 1382129_at - -
          Rat Expression Set 230 1382112_at - -
          Rat Expression Set 230 1385158_PM_at - -
          Rat Expression Set 230 1382112_PM_at - -
          Rat Expression Set 230 1385158_at - -
          Rat Expression Set 230 1382129_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802405 - -
          Rat Genome U34 Set rc_AA963240_at - -
          Rat Genome U34 Set rc_AI136136_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20969778 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline