Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01492020_m1
          See other RBM22 GE Assays ›
          Gene Symbol:
          Rbm22
          Gene Name:
          RNA binding motif protein 22
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 55828158 - 55838966 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01492020_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025676.1 NP_001020847.1 8 - 9 967 80
          GenBank mRNA BC097991.1 - 8 - 9 967 7446014 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000612.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm22

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          positive regulation of RNA splicing
          cellular response to drug
          mRNA cis splicing, via spliceosome
          positive regulation of intracellular protein transport
          nucleotide binding
          nucleocytoplasmic transporter activity
          snRNA binding
          pre-mRNA binding
          poly(A) RNA binding
          metal ion binding
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P241324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107763 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P241324 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P241324 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P241324 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P241324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90866 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802004 - -
          Affymetrix Rat Gene 1.0 ST Array 10802006 - -
          Affymetrix Rat Gene 1.0 ST Array 10802000 - -
          Affymetrix Rat Gene 1.0 ST Array 10802001 - -
          Affymetrix Rat Gene 1.0 ST Array 10802003 - -
          Affymetrix Rat Gene 1.0 ST Array 10802005 - -
          Affymetrix Rat Gene 1.0 ST Array 10801998 - -
          Affymetrix Rat Gene 1.0 ST Array 10802002 - -
          Affymetrix Rat Gene 1.0 ST Array 10801997 - -
          Affymetrix Rat Gene 1.0 ST Array 10801999 - -
          Affymetrix Rat Gene 1.1 ST Array 10801996 - -
          Rat Genome 230 2.0 Array 1392503_PM_at - -
          Rat Genome 230 2.0 Array 1392503_at - -
          Affymetrix HT Rat Focus Array 1392503_at - -
          Affymetrix HT Rat Focus Array 1392503_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800004443.rn.1 TC1800000612.rn.1 RTA TC
          Rat Expression Set 230 1392503_PM_at - -
          Rat Expression Set 230 1392503_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801996 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21624515 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline