Search
Search
| Gene Symbol: Dcp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749821.3 | XP_003749869.1 | 4 - 5 | 827 | 71 | |
| XM_008763272.1 | XP_008761494.1 | 4 - 5 | 827 | 71 | ||
| XM_575654.6 | XP_575654.3 | 4 - 5 | 827 | 71 | ||
Set Membership: |
|