Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01492482_m1
          See other USP18 GE Assays ›
          Gene Symbol:
          Usp18
          Gene Name:
          ubiquitin specific peptidase 18
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 153812312 - 153834374 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01492482_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usp18
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014058.1 NP_001014080.1 9 - 10 1160 87
          GenBank mRNA BC079101.1 - 9 - 10 1160 7124857 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001709.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Usp18

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein deubiquitination
          ISG15-protein conjugation
          response to stilbenoid
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of proteasomal protein catabolic process
          cysteine-type endopeptidase activity
          thiol-dependent ubiquitin-specific protease activity
          ISG15-specific protease activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P157059 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P284200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P284200 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P157059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P284200 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P157059 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P284200 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P157229 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P284200 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65322 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858373 - -
          Affymetrix Rat Gene 1.0 ST Array 10858371 - -
          Affymetrix Rat Gene 1.0 ST Array 10858372 - -
          Affymetrix Rat Gene 1.1 ST Array 10858370 - -
          Rat Genome 230 2.0 Array 1389034_PM_at - -
          Rat Genome 230 2.0 Array 1389034_at - -
          Affymetrix HT Rat Focus Array 1389034_PM_at - -
          Affymetrix HT Rat Focus Array 1389034_at - -
          Affymetrix Rat Clariom S Assay PSR0400012178.rn.1 TC0400001709.rn.1 RTA TC
          Rat Expression Set 230 1389034_PM_at - -
          Rat Expression Set 230 1389034_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858370 - -
          Rat Genome U34 Set rc_AA851237_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21380200 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349649 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0