Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01492499_m1
          See other SLC6A13 GE Assays ›
          Gene Symbol:
          Slc6a13
          Gene Name:
          solute carrier family 6 (neurotransmitter transporter), member 13
          Gene Aliases:
          GAT-2
          Chromosome Location:
          Chr.4: 153874943 - 153912185 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01492499_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc6a13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133623.1 NP_598307.1 9 - 10 1185 62
          XM_006237218.2 XP_006237280.1 9 - 10 1187 62
          XM_006237219.2 XP_006237281.1 10 - 11 1303 62
          XM_006237220.2 XP_006237282.1 9 - 10 1169 62
          XM_006237221.2 XP_006237283.1 10 - 11 1285 62
          XM_006237222.2 XP_006237284.1 8 - 9 1041 62
          XM_006237223.2 XP_006237285.1 8 - 9 1168 62
          GenBank mRNA M95762.1 - 9 - 10 1185 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001710.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Transporter Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc6a13

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter transport
          gamma-aminobutyric acid transport
          transmembrane transport
          neurotransmitter:sodium symporter activity
          gamma-aminobutyric acid:sodium symporter activity
          neurotransmitter binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P313272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23605 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137054 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P313272 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P313272 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P313272 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P313272 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700490925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20423 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8088 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10374 - -
          Agilent Rat cDNA Microarray (G4105A) 700490925 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858386 - -
          Affymetrix Rat Gene 1.0 ST Array 10858382 - -
          Affymetrix Rat Gene 1.0 ST Array 10858385 - -
          Affymetrix Rat Gene 1.0 ST Array 10858387 - -
          Affymetrix Rat Gene 1.0 ST Array 10858383 - -
          Affymetrix Rat Gene 1.0 ST Array 10858380 - -
          Affymetrix Rat Gene 1.0 ST Array 10858381 - -
          Affymetrix Rat Gene 1.0 ST Array 10858389 - -
          Affymetrix Rat Gene 1.0 ST Array 10858384 - -
          Affymetrix Rat Gene 1.0 ST Array 10858388 - -
          Affymetrix Rat Gene 1.0 ST Array 10858376 - -
          Affymetrix Rat Gene 1.0 ST Array 10858378 - -
          Affymetrix Rat Gene 1.0 ST Array 10858379 - -
          Affymetrix Rat Gene 1.0 ST Array 10858375 - -
          Affymetrix Rat Gene 1.1 ST Array 10858374 - -
          Rat Genome 230 2.0 Array 1387372_at - -
          Rat Genome 230 2.0 Array 1387372_PM_at - -
          Affymetrix HT Rat Focus Array 1387372_at - -
          Affymetrix HT Rat Focus Array 1387372_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400012194.rn.1 TC0400001710.rn.1 RTA TC
          Rat Expression Set 230 1387372_PM_at - -
          Rat Expression Set 230 1387372_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858374 - -
          Rat Genome U34 Set M95762_at - -
          Rat Genome U34 Set M95762_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351129 - -
          RatRef-12 Expression BeadChip ILMN_64063 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline