Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033441.1 | NP_254276.1 | 4 - 5 | 1015 | 96 | |
GenBank mRNA | Z46957.1 | - | 4 - 5 | 1015 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons |