Search
Search

B4galnt3| Gene Symbol: B4galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191976.1 | NP_001178905.1 | 17 - 18 | 2585 | 65 | |
| XM_006237267.2 | XP_006237329.1 | 17 - 18 | 2888 | 65 | ||
Set Membership: |
|