Search
Search

Plekha5| Gene Symbol: Plekha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763357.1 | XP_008761579.1 | 2 - 3 | 256 | 81 | |
| XM_008763358.1 | XP_008761580.1 | 2 - 3 | 256 | 81 | ||
| XM_008763359.1 | XP_008761581.1 | 2 - 3 | 256 | 81 | ||
| XM_008763360.1 | XP_008761582.1 | 2 - 3 | 256 | 81 | ||
| XM_008763362.1 | XP_008761584.1 | 2 - 3 | 256 | 81 | ||
| XM_008763364.1 | XP_008761586.1 | 2 - 3 | 256 | 81 | ||
| XM_008763365.1 | XP_008761587.1 | 2 - 3 | 256 | 81 | ||
| XM_008763366.1 | XP_008761588.1 | 2 - 3 | 256 | 81 | ||
| XM_008763367.1 | XP_008761589.1 | 2 - 3 | 256 | 81 | ||
Set Membership: |
|