Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493027_m1
          See other IL17RA GE Assays ›
          Gene Symbol:
          Il17ra
          Gene Name:
          interleukin 17 receptor A
          Gene Aliases:
          Il17r
          Chromosome Location:
          Chr.4: 152995865 - 153018394 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493027_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il17ra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107883.2 NP_001101353.2 3 - 4 411 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400001683.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          positive regulation of interleukin-23 production
          cellular response to cytokine stimulus
          fibroblast activation
          positive regulation of cytokine production involved in inflammatory response
          molecular_function
          interleukin-17 receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P046853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136192 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046853 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P046853 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P046853 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21070 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P129187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10858323 - -
          Affymetrix Rat Gene 1.0 ST Array 10858320 - -
          Affymetrix Rat Gene 1.0 ST Array 10858318 - -
          Affymetrix Rat Gene 1.0 ST Array 10858322 - -
          Affymetrix Rat Gene 1.0 ST Array 10858325 - -
          Affymetrix Rat Gene 1.0 ST Array 10858326 - -
          Affymetrix Rat Gene 1.0 ST Array 10858317 - -
          Affymetrix Rat Gene 1.0 ST Array 10858316 - -
          Affymetrix Rat Gene 1.0 ST Array 10858327 - -
          Affymetrix Rat Gene 1.0 ST Array 10858321 - -
          Affymetrix Rat Gene 1.0 ST Array 10858319 - -
          Affymetrix Rat Gene 1.1 ST Array 10858315 - -
          Rat Genome 230 2.0 Array 1373611_at - -
          Rat Genome 230 2.0 Array 1373611_PM_at - -
          Affymetrix HT Rat Focus Array 1373611_PM_at - -
          Affymetrix HT Rat Focus Array 1373611_at - -
          Affymetrix Rat Clariom S Assay PSR0400012042.rn.1 TC0400001683.rn.1 RTA TC
          Rat Expression Set 230 1373611_PM_at - -
          Rat Expression Set 230 1373611_at - -
          Affymetrix Rat Gene 1.0 ST Array 10858315 - -
          Rat Genome U34 Set rc_AA900972_at - -
          Rat Genome U34 Set rc_AA851044_s_at - -
          Rat Genome U34 Set rc_AI177312_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356655 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline