Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493384_m1
          See other LARP1B GE Assays ›
          Gene Symbol:
          Larp1b
          Gene Name:
          La ribonucleoprotein domain family, member 1B
          Gene Aliases:
          RGD1307509
          Chromosome Location:
          Chr.2: 127845034 - 127876793 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493384_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Larp1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014036.1 NP_001014058.1 8 - 9 948 129
          GenBank mRNA BC081895.1 - 8 - 9 948 7110249 129

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001050.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Larp1b

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P764792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67992 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106448 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P764792 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P764792 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P764792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83564 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10815155 - -
          Affymetrix Rat Gene 1.0 ST Array 10815157 - -
          Affymetrix Rat Gene 1.0 ST Array 10815154 - -
          Affymetrix Rat Gene 1.0 ST Array 10815156 - -
          Affymetrix Rat Gene 1.0 ST Array 10815152 - -
          Affymetrix Rat Gene 1.0 ST Array 10815158 - -
          Affymetrix Rat Gene 1.0 ST Array 10815153 - -
          Affymetrix Rat Gene 1.0 ST Array 10815161 - -
          Affymetrix Rat Gene 1.0 ST Array 10815159 - -
          Affymetrix Rat Gene 1.0 ST Array 10815151 - -
          Affymetrix Rat Gene 1.1 ST Array 10815150 - -
          Rat Genome 230 2.0 Array 1375222_at - -
          Rat Genome 230 2.0 Array 1375222_PM_at - -
          Rat Genome 230 2.0 Array 1379473_PM_at - -
          Rat Genome 230 2.0 Array 1379473_at - -
          Rat Genome 230 2.0 Array 1382219_PM_at - -
          Rat Genome 230 2.0 Array 1379044_PM_at - -
          Rat Genome 230 2.0 Array 1379044_at - -
          Rat Genome 230 2.0 Array 1382219_at - -
          Affymetrix HT Rat Focus Array 1379044_PM_at - -
          Affymetrix HT Rat Focus Array 1379044_at - -
          Affymetrix HT Rat Focus Array 1379473_at - -
          Affymetrix HT Rat Focus Array 1382219_at - -
          Affymetrix HT Rat Focus Array 1382219_PM_at - -
          Affymetrix HT Rat Focus Array 1379473_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200006731.rn.1 TC0200001050.rn.1 RTA TC
          Rat Expression Set 230 1379044_at - -
          Rat Expression Set 230 1379044_PM_at - -
          Rat Expression Set 230 1382219_PM_at - -
          Rat Expression Set 230 1382219_at - -
          Rat Expression Set 230 1379473_at - -
          Rat Expression Set 230 1379473_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10815150 - -
          Rat Genome U34 Set rc_AI639306_at - -
          Rat Genome U34 Set rc_AA945904_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354550 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline