Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493504_m1
          See other PRKCI GE Assays ›
          Gene Symbol:
          Prkci
          Gene Name:
          protein kinase C, iota
          Gene Aliases:
          Pkcl
          Chromosome Location:
          Chr.2: 115941998 - 116002550 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493504_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkci
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032059.1 NP_114448.1 6 - 7 733 68
          GenBank mRNA EU517502.1 - 6 - 7 565 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003421.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Apoptosis Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          actin filament organization
          positive regulation of neuron projection development
          cell migration
          cellular response to insulin stimulus
          negative regulation of glial cell apoptotic process
          cellular protein localization
          establishment of apical/basal cell polarity
          intracellular signal transduction
          eye photoreceptor cell development
          response to peptide hormone
          negative regulation of neuron apoptotic process
          cell-cell junction organization
          positive regulation of glucose import
          Golgi vesicle budding
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of glial cell proliferation
          response to interleukin-1
          positive regulation of establishment of protein localization to plasma membrane
          positive regulation of endothelial cell apoptotic process
          molecular_function
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          ATP binding
          phospholipid binding
          protein domain specific binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019643 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P602880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125767 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96861 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164006 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34311 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42277 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019643 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P602880 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P602880 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019643 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019643 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P602880 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557344 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P557345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90260 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14652 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822659 - -
          Affymetrix Rat Gene 1.0 ST Array 10822645 - -
          Affymetrix Rat Gene 1.0 ST Array 10822650 - -
          Affymetrix Rat Gene 1.0 ST Array 10822651 - -
          Affymetrix Rat Gene 1.0 ST Array 10822658 - -
          Affymetrix Rat Gene 1.0 ST Array 10822648 - -
          Affymetrix Rat Gene 1.0 ST Array 10822656 - -
          Affymetrix Rat Gene 1.0 ST Array 10822657 - -
          Affymetrix Rat Gene 1.0 ST Array 10822662 - -
          Affymetrix Rat Gene 1.0 ST Array 10822661 - -
          Affymetrix Rat Gene 1.0 ST Array 10822647 - -
          Affymetrix Rat Gene 1.0 ST Array 10822652 - -
          Affymetrix Rat Gene 1.0 ST Array 10822649 - -
          Affymetrix Rat Gene 1.0 ST Array 10822654 - -
          Affymetrix Rat Gene 1.0 ST Array 10822660 - -
          Affymetrix Rat Gene 1.0 ST Array 10822646 - -
          Affymetrix Rat Gene 1.0 ST Array 10822653 - -
          Affymetrix Rat Gene 1.0 ST Array 10822655 - -
          Affymetrix Rat Gene 1.1 ST Array 10822644 - -
          Rat Genome 230 2.0 Array 1371203_PM_at - -
          Rat Genome 230 2.0 Array 1373047_PM_at - -
          Rat Genome 230 2.0 Array 1373047_at - -
          Rat Genome 230 2.0 Array 1388251_at - -
          Rat Genome 230 2.0 Array 1371203_at - -
          Rat Genome 230 2.0 Array 1388251_PM_at - -
          Affymetrix HT Rat Focus Array 1373047_at - -
          Affymetrix HT Rat Focus Array 1388251_at - -
          Affymetrix HT Rat Focus Array 1371203_PM_at - -
          Affymetrix HT Rat Focus Array 1371203_at - -
          Affymetrix HT Rat Focus Array 1373047_PM_at - -
          Affymetrix HT Rat Focus Array 1388251_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200023331.rn.1 TC0200003421.rn.1 RTA TC
          Rat Expression Set 230 1371203_PM_at - -
          Rat Expression Set 230 1388251_PM_at - -
          Rat Expression Set 230 1373047_at - -
          Rat Expression Set 230 1388251_at - -
          Rat Expression Set 230 1371203_at - -
          Rat Expression Set 230 1373047_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822644 - -
          Rat Genome U34 Set rc_AI179206_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21905413 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369025 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline